DL-Limonen , tech.

Lieferant: Thermo Scientific
Achtung

Synonyme: (±)-Limonen, (±)-4-Isopropenyl-1-methyl-1-cyclohexen, (±)-Dipenten, (±)-p-Mentha-1,8-dien, (±)-1-Methyl-4-(1-methylvinyl)cyclohexen

45805.AK
45805.AKEA 50.2 EUR
45805.AK
DL-Limonen , tech.
(±)-p-Mentha-1,8-dien

CAS No.: 138-86-3 Molecular Formula: C10H16 Formula Weight: 136.24 Flash Point: 43°(109°F) Physical Form: Liquid Density: 0.85 Refractive Index: 1.474 Boiling Point: 170-180° Beilstein Reference: 3587825 Merck Reference: 14,5493 UN No.: 2052 MDL No.: MFCD00062992

Formel: C₁₀H₁₆
Molecular Weight: 136,24 g/mol
Siedepunkt: 170…180 °C
Schmelzpunkt: –89 °C
Dichte: 0,85
Flammpunkt: 43 °C (109 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00062992
CAS-Nummer: 138-86-3
EINECS: 205-341-0
UN: 2052
ADR: 3,III
Merck Index: 14,5493

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR