Zeolite Y, hydrogen

Lieferant: Thermo Fisher Scientific

45868.14 45868.22 45868.36
45868.14EA 40 EUR
45868.14 45868.22 45868.36
Zeolite Y, hydrogen
Zeolite Y, hydrogen

CAS No.: 1318-02-1 Molecular Formula: SiO2:Al2O3 Physical Form: Powder, S.A. 660 m2/g, 5.2:1 mole ratio Appearance: White MDL No.: MFCD00132601


Applications: Zeolite Y is used as a cracking catalyst in petroleum refinery catalytic cracking units to convert high-boiling fractions of petroleum crude to more valuable gasoline, diesel and other products

Formel: SiO₂:Al₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00132601
CAS-Nummer: 1318-02-1
EINECS: 215-283-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR