Zeolite mordenite, sodium

Lieferant: Thermo Fisher Scientific

45876.14 45876.22 45876.36
45876.14EA 44.6 EUR
45876.14 45876.22 45876.36
Zeolite mordenite, sodium
Zeolite mordenite, sodium

CAS No.: 1318-02-1 Molecular Formula: SiO2:Al2O3 Physical Form: Powder, S.A. 425 m2/g, 13:1 mole ratio Appearance: White MDL No.: MFCD00132601


Applications: Zeolite mordenite is used for the acid-catalyzed isomerization of alkanes and aromatics

Formel: SiO₂:Al₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00132601
CAS-Nummer: 1318-02-1
EINECS: 215-283-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR