Allura red AC 80% (Farbstoffgehalt), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: C.I. Food Red 17, Red No. 40, FD and C Red No.40 (FD&C Red 40), Rot Allura, C. I. 16035

M-Clarity™ Qualitätsstufe = MQ200

458848-100G 458848-500G
458848-100GEA 85.5 EUR
458848-100G 458848-500G
Allura red AC 80% (Farbstoffgehalt), Sigma-Aldrich®
Allura red AC

Allura Red AC is a food azo dye. It is a dark red powder or granules, that is soluble in water and insoluble in ethanol. Allura Red AC has been used: for the determination of fecal neutral sterols (FNS) in mice, to evaluate its developmental toxicity, as a coloring reagent for observing the experimental performance of liquid-handling robot.

Formel: C₁₈H₁₄N₂O₈S₂·2Na
Siedepunkt: 310 °C at 966 hPa
Schmelzpunkt: >300 °C (Lit.)
Dichte: 0,448 g/cm³ (20 °C, 976,8 hPa)
Lagertemperatur: Room Temperature
MDL: MFCD00059526
CAS-Nummer: 25956-17-6
EINECS: 247-368-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=078aaa0023281f2ca5462712123965304c943059&vid=078aaa0023281f2ca5462712123965304c943059&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Colour): Red to Maroon Conforms to Requirements
Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Wavelength, c = 0,01 g/L; Water 501 - 507 nm
Extinction Coefficient ≥20000
Carbon ≥34.0%
Nitrogen ≥4.4%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR