Incineration dishes, numbered for identification
Lieferant: MORGAN ADVANCED MATERIALS HALD
L33/2NUMBAB100
459-2228EA 0 EUR
459-2228
Incineration dishes, numbered for identification
Schiffchen Verbrennungsschiffchen
Porcelain.
  • Glazed inside and out, except base
  • Good chemical resistance
  • Withstand maximum temperature of 1000 °C
Numbers 1 to 99 or 100 to 999 are printed prior to glazing so that the identification of dishes is simple and not affected by the high temperatures of incineration.
Bestellinformation: At time of order, please request the identification number required.