Okadasäure Ammoniumsalz, Millipore®

Lieferant: Merck Millipore Calbiochem

Synonyme: Halochondrine A ammonium salt, OA

459616-25
459616-25EA 101 EUR
459616-25
Okadasäure Ammoniumsalz, Millipore®
Okadaic acid ammonium salt
Formel: C₄₄H₆₇O₁₃·NH₄
Molecular Weight: 822,03 g/mol
MDL: MFCD00153856
CAS-Nummer: 155716-06-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR