Okadasäure Kaliumsalz, Millipore®
Lieferant: Merck Millipore Calbiochem
Synonyme:
Halochondrine A potassium salt, OA potassium salt, 9,10-deepithio-9,10-didehydroacanthifolicin potassium salt
459618-50
459618-50EA
190
EUR
459618-50
Okadasäure Kaliumsalz, Millipore®
Okadaic acid potassium salt
Formel:
C₄₄H₆₇O₁₃·K Molecular Weight: 843.11 g/mol |
MDL:
MFCD00214356 CAS-Nummer: 155751-72-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...