Fenclorim, Supelco®

Lieferant: Merck

Synonyme: 4,6-Dichloro-2-phenylpyrimidine

M-Clarity™ Qualitätsstufe = MQ100

46005-250MG
46005-250MGEA 117 EUR
46005-250MG
Fenclorim, Supelco®
Fenclorim
Formel: C₁₀H₆Cl₂N₂
MDL: MFCD00754193
CAS-Nummer: 3740-92-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba5bf6301767952b638ac41bb3f630d052d70b02&vid=7208d9a46f69161c1809c08b9e09e00c6bd6c340&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR