[12-(4-Ethylphenyl)dodecyl] 2-nitrophenyl ether ≥95,0%, Selectophore™ Ionophor, Supelco®

Lieferant: Merck

Synonyme: ETH 8045

M-Clarity™ Qualitätsstufe = MQ100

46092-250MG-F
46092-250MG-FEA 177 EUR
46092-250MG-F
[12-(4-Ethylphenyl)dodecyl] 2-nitrophenyl ether ≥95,0%, Selectophore™ Ionophor, Supelco®
[12-(4-Ethylphenyl)dodecyl] 2-nitrophenyl ether

[12-(4-Ethylphenyl)dodecyl] 2-nitrophenyl ether is a plasticizer mainly used during electrode preparation.


It was used in preparation of PVC membrane cocktail with multi-wall carbon nanotubes, in a study done to investigate the use of solid-contact (SC) ion-selective electrodes (ISE) in combination with an radio-frequency identification potentiometer.


Lipophilic plasticizer (log PTLC=12.8) with excellent characteristics for use in magnesium selective membrane electrodes.


Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.

Formel: C₂₆H₃₇NO₃
Molecular Weight: 411,58 g/mol
MDL: MFCD00216702
CAS-Nummer: 155056-63-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e23103ba0a687a72aea03bc12fb3278f0385a067&vid=677d6dce8407372485fccfdfcb95f9334b23f856&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR