VWR®, Fern-Temperaturfühler für VWR Kühler
Lieferant: VWR Collection
461-0101EA 411 EUR
461-0101 461-0102 461-0103
VWR®, Fern-Temperaturfühler für VWR Kühler
Temperaturfühler
Remote temperature probes for circulator use with a chiller. Allows points of control to be switched from inside a chiller to a remote location. Compensates for heat loss through tubing between the circulator and the control point. Platinum RTD sensor with stainless steel sheath. Rubber coupling with 25,4 mm Ø, connects sheath to flexible cable. Probe cable ends in a DB9 female plug.
Länge×Ø: 152×4 mm
Bestellen
TECHNISCHE DATEN
Environmentally Preferable