Dapson, Supelco®
Lieferant: Merck
|
Achtung
|
Synonyme:
DDS, 4-Aminophenylsulfon, Bis(4-aminophenyl)sulfon, 4,4'-Sulfonyldianilin, 4,4'-Diaminodiphenylsulfon
M-Clarity™ Qualitätsstufe = MQ200
46158-250MG
46158-250MGEA
46.5
EUR
46158-250MG
Dapson, Supelco®
Dapson
Formel:
C₁₂H₁₂N₂O₂S |
MDL:
MFCD00007887 CAS-Nummer: 80-08-0 EINECS: 201-248-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...