Dexamethason, Supelco®

Lieferant: Merck

Synonyme: 9-Fluor-11β,17α,21-trihydroxy-16α-methyl-pregna-1,4-dien-3,20-dion, Dexamethasonum

M-Clarity™ Qualitätsstufe = MQ100

46165-250MG
46165-250MGEA 101 EUR
46165-250MG
Dexamethason, Supelco®
Dexamethason
Formel: C₂₂H₂₉FO₅
Molecular Weight: 392,47 g/mol
Schmelzpunkt: 268…271 °C
MDL: MFCD00064136
CAS-Nummer: 50-02-2
EINECS: 200-003-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da8555814cb05b7746c78831a8328900775cdd54&vid=0eb6e28b0a8607228aedf6fd3ddbd58281e6287c&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR