VWR® MX, Eintauchthermostat
Lieferant: VWR Collection
462-0205EA 2190 EUR
462-0205
VWR® MX, Eintauchthermostat
Umwälzthermostate Offene Umwälzsysteme Thermostatische Regler
The MX immersion thermostat is user friendly with three control buttons and on-screen prompts. The convenient slide control enables the flow rate to be easily adjusted, and it can clamp securely to straight or curved tank walls.
  • Large EasyView™ LCD display with alarm or fault icons plus English prompts and menus
  • User-adjustable high and low temperature limits and low liquid level alarms
  • Single speed pressure pump
  • Integral pump and heater coil protection
  • Single point calibration
Maximum pump capacities:
11,9 l/min, 120 mbar.
Maximum pump capacities:
10,6 l/min, 100 mbar
Standards: Complies with DIN 12876-1, Safety Class I.
Bitte beachten * RT = Ambient.