VWR®, External PT100 Probe for Chillers
Lieferant: VWR Collection
462-0553EA 445 EUR
462-0553
VWR®, External PT100 Probe for Chillers
Temperaturfühler
External PT100 probe is used for with chillers and coolers, 6000 series chiller, non-refrigerated coolers, 4100 liquid-to-liquid coolers.