Methyl-α-D-mannopyranosid  ≥99% (durch TLC), Calbiochem®, Sigma-Aldrich®

Lieferant: Merck Millipore Calbiochem

Synonyme: ɑ-Methyl-D-mannoside, Methyl-ɑ-D-Mannoside, Methyl α-D-Mannopyranoside, ɑ-Methyl-D-mannopyranoside, (2R,3S,4S,5S,6S)-2-(Hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3,4,5-triol

462711-100
462711-100EA 143 EUR
462711-100
Methyl-α-D-mannopyranosid  ≥99% (durch TLC), Calbiochem®, Sigma-Aldrich®
Methyl-α-D-mannopyranosid 

Methyl-α-D-mannopyranoside is a substrate for α1,2-mannosyltransferase.

Formel: C₇H₁₄O₆
Molecular Weight: 194,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00063262
CAS-Nummer: 617-04-9

Bestellen

Spezifikation Testergebnisse

Purity (by TLC) ≥99%
Heavy Metals ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR