Ammoniumsulfat 99,0-100,5% NF, ACS, J.T.Baker®

Lieferant: Avantor
Achtung

Synonyme: Diammoniumsulfat, di-Ammoniumsulfat

Veränderungsmanagement-Kategorie= R

4628-01EA 0 EUR
4628-01 4628-F9
Ammoniumsulfat 99,0-100,5% NF, ACS, J.T.Baker®
Ammoniumsulfat

  • GMP manufactured product

Formel: (NH₄)₂SO₄
Molecular Weight: 132.14 g/mol
MDL: MFCD00003391
CAS-Nummer: 7783-20-2
Merck Index: 13,00559

Bestellen

Spezifikation Testergebnisse

GMP Manufactured Product 99.0 - 100.5%
Meets N.F. Requirements ≤0.005%
Meets ACS Reagent Chemical Requirements ≤0.005%
CAUTION: For Manufacturing, processing or repackaging 5.0 - 6.0
Bulk Pharmaceutical Chemical ≤0.001%
NF - Assay ((NH₄)₂SO₄) 99.0 - 100.5 %
NF - Insoluble Matter ≤ 0.005 %
NF - Residue on Ignition ≤ 0.005 %
NF - pH of 5% Solution 5.0 - 6.0
NF - Nitrate (NO₃) ≤ 0.001 %
NF - Chloride (Cl) ≤ 5 ppm
NF - Phosphate (PO₄) ≤ 5 ppm
NF - Iron (Fe) ≤ 5 ppm
NF - Identification Passes Test
NF - Microbial Limit - Total Aerobic Microbial Count (cfu/g) ≤ 1000
NF - Microbial Limit - Total Combined Molds and Yeasts (cfu/g) ≤ 10
Endotoxin Concentration (EU/g) ≤ 2.5
Nickel (Ni) ≤ 0.7 ppm
Cobalt (Co) ≤ 0.09 ppm
Chromium (Cr) ≤ 0.75 ppm
Copper (Cu) ≤ 0.03 ppm
Manganese (Mn) ≤ 3.45 ppm
Zinc (Zn) (For Information Only) ≤5 ppm
Ni / Zn Ratio (For Information Only) ≤5 ppm
Co / Zn Ratio (For Information Only) ≤5 ppm
Cu / Zn Ratio (For Information Only) ≥99.0 %
ACS - Assay ((NH₄)₂SO₄) (A.C.S.) ≥ 99.0 %
ACS - Residue on Ignition ≤ 0.005 %
ACS - pH of 5% Solution at 25°C 5.0 - 6.0
ACS - Insoluble Matter ≤ 0.005 %
ACS - Chloride (Cl) ≤ 5 ppm
ACS - Nitrate (NO₃) ≤ 0.001 %
ACS - Phosphate (PO₄) ≤ 5 ppm
ACS - Heavy Metals (as Pb) ≤ 5 ppm
ACS - Iron (Fe) ≤ 5 ppm
Solution Test Passes Test
No Class 1, 2 or 3 Residual Solvents are likely to be present by
exclusion of the production/purification process.
Elemental Impurities (USP (232), EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.
Preserve in well closed container.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR