Ammoniumsulfat 99,0-100,5% NF, ACS, J.T.Baker®
Lieferant: Avantor
|
Achtung
|
Synonyme:
Diammoniumsulfat, di-Ammoniumsulfat
Veränderungsmanagement-Kategorie= R
4628-01EA
0
EUR
4628-01
4628-F9
Ammoniumsulfat 99,0-100,5% NF, ACS, J.T.Baker®
Ammoniumsulfat
- GMP manufactured product
Formel:
(NH₄)₂SO₄ Molecular Weight: 132.14 g/mol |
MDL:
MFCD00003391 CAS-Nummer: 7783-20-2 Merck Index: 13,00559 |
Spezifikation Testergebnisse
GMP Manufactured Product | 99.0 - 100.5% |
Meets N.F. Requirements | ≤0.005% |
Meets ACS Reagent Chemical Requirements | ≤0.005% |
CAUTION: For Manufacturing, processing or repackaging | 5.0 - 6.0 |
Bulk Pharmaceutical Chemical | ≤0.001% |
NF - Assay ((NH₄)₂SO₄) | 99.0 - 100.5 % |
NF - Insoluble Matter | ≤ 0.005 % |
NF - Residue on Ignition | ≤ 0.005 % |
NF - pH of 5% Solution | 5.0 - 6.0 |
NF - Nitrate (NO₃) | ≤ 0.001 % |
NF - Chloride (Cl) | ≤ 5 ppm |
NF - Phosphate (PO₄) | ≤ 5 ppm |
NF - Iron (Fe) | ≤ 5 ppm |
NF - Identification | Passes Test |
NF - Microbial Limit - Total Aerobic Microbial Count (cfu/g) | ≤ 1000 |
NF - Microbial Limit - Total Combined Molds and Yeasts (cfu/g) | ≤ 10 |
Endotoxin Concentration (EU/g) | ≤ 2.5 |
Nickel (Ni) | ≤ 0.7 ppm |
Cobalt (Co) | ≤ 0.09 ppm |
Chromium (Cr) | ≤ 0.75 ppm |
Copper (Cu) | ≤ 0.03 ppm |
Manganese (Mn) | ≤ 3.45 ppm |
Zinc (Zn) (For Information Only) | ≤5 ppm |
Ni / Zn Ratio (For Information Only) | ≤5 ppm |
Co / Zn Ratio (For Information Only) | ≤5 ppm |
Cu / Zn Ratio (For Information Only) | ≥99.0 % |
ACS - Assay ((NH₄)₂SO₄) (A.C.S.) | ≥ 99.0 % |
ACS - Residue on Ignition | ≤ 0.005 % |
ACS - pH of 5% Solution at 25°C | 5.0 - 6.0 |
ACS - Insoluble Matter | ≤ 0.005 % |
ACS - Chloride (Cl) | ≤ 5 ppm |
ACS - Nitrate (NO₃) | ≤ 0.001 % |
ACS - Phosphate (PO₄) | ≤ 5 ppm |
ACS - Heavy Metals (as Pb) | ≤ 5 ppm |
ACS - Iron (Fe) | ≤ 5 ppm |
Solution Test | Passes Test |
No Class 1, 2 or 3 Residual Solvents are likely to be present by | |
exclusion of the production/purification process. | |
Elemental Impurities (USP (232), EP 5.20) - Information on elemental | |
impurities for this product is available on the associated Product | |
Regulatory Data Sheet and elemental impurity profile report. | |
Preserve in well closed container. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...