Kobalt ≥99,8% (Metall-Basis), Nano-Pulver APS 25-30 nm

Lieferant: Thermo Fisher Scientific
Gefahr

46347.06
46347.06EA 130 EUR
46347.06
Kobalt ≥99,8% (Metall-Basis), Nano-Pulver APS 25-30 nm
Cobalt
Formel: Co
Lagertemperatur: Raumtemperatur
MDL: MFCD00010935
CAS-Nummer: 7440-48-4
EINECS: 231-158-0
UN: 3089
ADR: 4.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR