Kristallviolett, Supelco®

Lieferant: Merck
Gefahr

Synonyme: Hexamethyl-p-rosaniliniumchlorid, Hexamethylen-p-rosanilinchlorid, blaues Pyoktanin, Basic Violet 3, Hexamethylviolett, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]dimethylammoniumchlorid, Methylviolett 10B, Methylrosanilin, Methylrosanilium, 4,4',4''-Tri(dimethylamino)trityliumchlorid, CI 42555, Methylrosaniliumchlorid, Gentianaviolett

M-Clarity™ Qualitätsstufe = MQ100

46364-250MG
46364-250MGEA 75.3 EUR
46364-250MG
Kristallviolett, Supelco®
Kristallviolett
Formel: C₂₅H₃₀ClN₃
Molecular Weight: 407,99 g/mol
Schmelzpunkt: 189…194 °C
Dichte: 1,19 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011750
CAS-Nummer: 548-62-9
EINECS: 208-953-6
UN: 3077
ADR: 9,III
Merck Index: 13,04406
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e373e953bce623fb18617f1c5a7deb3db4da356&vid=7c7c205485c85d82bb270c503cff84046fea47c5&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR