Diaion SK1B(Na), ion exchange resin, 8% cross-linked, strongly acidic gel type, 2.0 meq/ml on PS-DVB 1 * 1 kg
Lieferant: Thermo Fisher Scientific
Diaion SK1B(Na), ion exchange resin, 8% cross-linked, strongly acidic gel type, 2.0 meq/ml on PS-DVB 1 * 1 kg