Polyester methacrylate PEM-665

Lieferant: Thermo Fisher Scientific
Achtung

46408.22 46408.14
46408.22EA 712 EUR
46408.22 46408.14
Polyester methacrylate PEM-665
Polyester methacrylate

Storage Temperature: 2°C to 8°C Physical Form: Liquid Appearance: Pale yellow


Applications: Good solubility in both aliphatic and aromatic co-monomers

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR