Chloro(cyclopentadienyl)bis(triphenylphosphine)ruthenium(II) ethanol adduct

Lieferant: Thermo Fisher Scientific
Achtung

46430.03 46430.06
46430.03EA 184 EUR
46430.03 46430.06
Chloro(cyclopentadienyl)bis(triphenylphosphine)ruthenium(II) ethanol adduct
Chloro(cyclopentadienyl)bis(triphenylphosphine)ruthenium(II) ethanol adduct

Cyclopentadienylbis(triphenylphosphine)ruthenium Chloride is used as an efficient transfer hydrogenation catalyst. It is used in the formation of indoles, isoquinolines and quinolines.


Moisture Sensitive. Air Sensitive. Store under Argon.

Formel: C₄₃H₄₁P₂ClORu
Lagertemperatur: Raumtemperatur
MDL: MFCD00075004
CAS-Nummer: 32993-05-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR