Diaion UBK530(Na), ion exchange fractionation resin, 6% cross-linked, strongly acidic gel type, 1.6 meq/ml on PS-DVB 1 * 1 kg
Lieferant: Thermo Scientific
Diaion UBK530(Na), ion exchange fractionation resin, 6% cross-linked, strongly acidic gel type, 1.6 meq/ml on PS-DVB 1 * 1 kg