Estriol, Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

46565-100MG
46565-100MGEA 99.8 EUR
46565-100MG
Estriol, Supelco®
Estriol
Formel: C₁₈H₂₄O₃
MDL: MFCD00003691
CAS-Nummer: 50-27-1
EINECS: 200-022-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f07c7b0fa04470db643f3d9b98cfc4a4223093f8&vid=6d2c6927ed336d7cbc65bb13ada87b7d3f6e0dc7&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR