Oxytetracyclin Hydrochlorid, Supelco®

Lieferant: Merck

46598-250MG
46598-250MGEA 55.4 EUR
46598-250MG
Oxytetracyclin Hydrochlorid, Supelco®
Oxytetracyclin Hydrochlorid
Formel: C₂₂H₂₅ClN₂O₉
MDL: MFCD00135815
CAS-Nummer: 2058-46-0
EINECS: 218-161-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b71fa3e72267cfeb10824fafb74a9b1369cd3876&vid=b71fa3e72267cfeb10824fafb74a9b1369cd3876&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR