Drying and heating chambers with forced convection, FP series Avantgarde.Line
Lieferant: Binder
9010-0363 9010-0366 9010-0365 9010-0364
466-0653EA 3342 EUR
466-0653 466-0656 466-0655 466-0654
Drying and heating chambers with forced convection, FP series Avantgarde.Line
Öfen Standard-Trockenschränke
The Binder FP series of material test chambers with mechanical convection and program functions provide reliably short drying times and particularly fast heating – even for chambers under full loads.
  • High temperature accuracy thanks to APT.line™ technology
  • Controller with interval and real-time programming
  • Adjustable fan speed
  • Electromechanical control of the exhaust air flap
  • Class 2 independent adjustable temperature safety device (DIN 12880) and class 3.1 independent temperature safety device (DIN 12880) with visual/acoustic alarm available for selection
More Product Information