Ion exchange resins, Diaion® UBK550 (Na)
Lieferant: Thermo Scientific
46609.A1
46609.A1EA
112
EUR
46609.A1
Ion exchange resins, Diaion® UBK550 (Na)
Chromatographie-Sorbentien
HPLC-Sorbentien
These resins are ideal for use in chromatographic oligosaccharide sugar separations and sugar alcohol separations.