7-Fluorobenzo-2-oxa-13-diazole-4-sulphonic acid ammonium salt ≥98,5% (durch HPLC), LiChropur™ Derivatisierungsreagenz, für die HPLC, Supelco®
Lieferant: Merck
Synonyme:
Ammonium 7-fluoro-2,1,3-benzoxadiazole-4-sulphonate
M-Clarity™ Qualitätsstufe = MQ100
46640-25MG-F
46640-25MG-FEA
425
EUR
46640-25MG-F
7-Fluorobenzo-2-oxa-13-diazole-4-sulphonic acid ammonium salt ≥98,5% (durch HPLC), LiChropur™ Derivatisierungsreagenz, für die HPLC, Supelco®
7-Fluorobenzo-2-oxa-13-diazole-4-sulphonic acid ammonium salt
Fluorescent probe for thiols and pre-column labeling of biological thiols for HPLC
Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.
Formel:
C₆H₆FN₃O₄S Molecular Weight: 235,189 g/mol |
MDL:
MFCD02682109 CAS-Nummer: 84806-27-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...