Sodium bis(trifluoromethylsulphonyl)imide
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Bis(trifluoromethanesulphonyl)imide sodium salt, Sodium triflimide, Sodium trifluoromethanesulphonimide, Sodium Bis(trifluoromethanesulphonyl)imide
46640.MD
46640.03
46640.06
46640.MDEA
37.5
EUR
46640.MD
46640.03
46640.06
Sodium bis(trifluoromethylsulphonyl)imide
Sodium bis(trifluoromethylsulphonyl)imide
CAS No.: 91742-21-1
Molecular Formula: C2F6NNaO4S2
Formula Weight: 303.13
Physical Form: Solid
Sensitivity: Moisture Sensitive
Melting Point: 257-258°
UN No.: UN3261
MDL No.: MFCD14156021
Formel:
C₂F₆NNaO₄S₂ Molecular Weight: 303.13 g/mol Schmelzpunkt: 257…258 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD14156021 CAS-Nummer: 91742-21-1 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...