Silicium ≥99,999% (Metall-Basis), Pulver -325 mesh
Lieferant: Thermo Fisher Scientific
|
Achtung
|
46654.09
46654.18
46654.09EA
95.2
EUR
46654.09
46654.18
Silicium ≥99,999% (Metall-Basis), Pulver -325 mesh
Silicium
Purity: 99.999%
CAS No.: 7440-21-3
Odour: Odourless
UN No.: UN1346
MDL No.: MFCD00085311
Formel:
Si Lagertemperatur: Raumtemperatur |
MDL:
MFCD00085311 CAS-Nummer: 7440-21-3 EINECS: 231-130-8 UN: 1346 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...