Diaion RCP160M(H), ion exchange resin, highly cross-linked, strongly acidic highly porous type, 1.5 meq/ml on PS-DVB 1 * 1 kg
Lieferant: Thermo Scientific
Diaion RCP160M(H), ion exchange resin, highly cross-linked, strongly acidic highly porous type, 1.5 meq/ml on PS-DVB 1 * 1 kg