Ecosurf™ EH-9
Lieferant: Thermo Scientific
Synonyme: 2-Ethyl hexanol EoPo copolymer (9 EO), Alcohol (2EH) alkoxylate, Ethyl hexanol ethoxylated propoxylated
467361000
467361000.EA 40 EUR
467361000.
Ecosurf™ EH-9
Ecosurf™ EH-9
Bestellen
Spezifikation Testergebnisse
Appearance milky white to light yellow liquid
Color scale ≤150 APHA (40°C)
pH 5.0 to 7.5 (1% in water)
Water ≤0.5 %
Cloud point 62°C to 67°C (10% in water)