Tetrabutylammoniumdihydrogenphosphat ≥98%, nicht-UV

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Tetra-n-butylammonium phosphate monobasic, Tetrabutylammonium phosphate monobasic, Tetra-n-butylammonium dihydrogen phosphate, IPC-TBA-P, Tetra-n-butylammonium phosphate

46746.04 46746.09 46746.18
46746.04EA 81.9 EUR
46746.04 46746.09 46746.18
Tetrabutylammoniumdihydrogenphosphat ≥98%, nicht-UV
Tetrabutylammoniumdihydrogenphosphat

Tetra-n-butylammonium phosphate, is used in chemicals, pharmaceutical research, pharmaceutical intermediates, and reagents. Tetrabutylammonium phosphate can be used in organic synthesis. It is used as an Ion-associating reagent for HPLC.

Formel: C₁₆H₃₈NO₄P
Schmelzpunkt: 151…154 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00064526
CAS-Nummer: 5574-97-0
EINECS: 226-947-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR