Allyl(chloro)[1,2,3,4,5-pentaphenyl-1'-(di-tert-butylphosphino)ferrocene]palladium(II)
Lieferant: Thermo Scientific
Synonyme:
PdClAllyl(Qphos)
46747.03
46747.03EA
358
EUR
46747.03
Allyl(chloro)[1,2,3,4,5-pentaphenyl-1'-(di-tert-butylphosphino)ferrocene]palladium(II)
Allyl(chloro)[1,2,3,4,5-pentaphenyl-1'-(di-tert-butylphosphino)ferrocene]palladium(II)
Formel:
C₅₁H₅₂ClFePPd Molecular Weight: 893,666 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD25372546 CAS-Nummer: 1248656-98-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...