Cer(IV)oxid 25% in Wasser, kolloidale Dispersion, NanoArc® CE-6440
Lieferant: Thermo Fisher Scientific
Achtung
|
CAS No.: 1306-38-3
Molecular Formula: CeO2
Formula Weight: 172.12
Physical Form: 30nm APS for dry powder (typical)
Odour: Odourless
MDL No.: MFCD00010933
Suitable for use in ultrafine acid side polishing slurries
Formel:
CeO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010933 CAS-Nummer: 1306-38-3 EINECS: 215-150-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...