Atomoxetine hydrochloride
Lieferant: Thermo Scientific
Synonyme:
(R)-N-Methyl-3-(2-methylphenoxy)-3-phenylpropylamine Hydrochloride, (R)-Tomoxetine Hydrochloride, (R)-N-Methyl-3-phenyl-3-(o-tolyloxy)propan-1-amine hydrochloride
467680010
467680010.EA
97.5
EUR
467680010.
Atomoxetine hydrochloride
Atomoxetine hydrochloride
Formel:
C₁₇H₂₁NO Molecular Weight: 291,82 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD06410992 CAS-Nummer: 82248-59-7 |
Spezifikation Testergebnisse
Appearance | white to beige powder |
Infrared spectrum | Conforms |
HPLC | ≥98.5 % |
Melting point | 167°C to 171°C |
Loss on drying | ≤0.5 % (1 g, 105°C, 2 hr) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...