Pyrrolo[1,2-a]pyridine

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Azaindene, Indolizine

467730010 467732500
467730010.EA 382 EUR
467730010. 467732500.
Pyrrolo[1,2-a]pyridine
Pyrrolo[1,2-a]pyridine
Formel: C₈H₇N
Siedepunkt: 202°C (760mmHg)
Lagertemperatur: Gefrierschrank
MDL: MFCD00030189
CAS-Nummer: 274-40-8

Bestellen

Spezifikation Testergebnisse

Appearance white to green-grey crystalline powder
Infrared spectrum Conforms
GC ≥94 %
Melting point 70°C to 78°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR