Ferene disodium salt

Lieferant: Thermo Fisher Scientific
Achtung

467750010 467750050 467750250
467750010.EA 55.4 EUR
467750010. 467750050. 467750250.
Ferene disodium salt
Ferene disodium salt
Formel: C₁₆H₈N₄Na₂O₈S₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00040462
CAS-Nummer: 79551-14-7

Bestellen

Spezifikation Testergebnisse

Appearance white to dark yellow powder or crystals
Infrared spectrum Conforms
HPLC ≥97.5 %
Water ≤13 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR