Ferene disodium salt
Lieferant: Thermo Fisher Scientific
Achtung
Synonyme: Sodium 5,5'-(3-(pyridin-2-yl)-1,2,4-triazine-5,6-diyl)bis(furan-2-sulphonate), Ferene, 3-(2-Pyridyl)-5,6-di(2-furyl)-1,2,4-triazine-5′,5′′-disulphonic acid disodium salt, Disodium 5-[3-pyridin-2-yl-6-(5-sulphonatofuran-2-yl)-1,2,4-triazin-5-yl]furan-2-sulphonate
467750010 467750050 467750250
467750010.EA 55.4 EUR
467750010. 467750050. 467750250.
Ferene disodium salt
Ferene disodium salt
Formel: C₁₆H₈N₄Na₂O₈S₂
Molecular Weight: 494.37 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00040462
CAS-Nummer: 79551-14-7
Bestellen
Spezifikation Testergebnisse
Appearance white to dark yellow powder or crystals
Infrared spectrum Conforms
HPLC ≥97.5 %
Water ≤13 %