Sulfachlorpyridazin, Supelco®

Lieferant: Merck

Synonyme: 4-Amino-N-(6-chloropyridazin-3-yl)benzenesulphonamide

M-Clarity™ Qualitätsstufe = MQ200

46778-250MG
46778-250MGEA 52.8 EUR
46778-250MG
Sulfachlorpyridazin, Supelco®
Sulfachlorpyridazin
Formel: C₁₀H₉ClN₄O₂S
Molecular Weight: 284,7221 g/mol
MDL: MFCD00057371
CAS-Nummer: 80-32-0
EINECS: 201-269-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=355646e50f2544963d839d8fda00732c1a1670f1&vid=b27050addf366ba0ab73b4cc36979f093807559e&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR