Polyethylene glycol diacrylate (PEGDA), M.W. 8,000
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
46801.MD
46801.03
46801.06
46801.MDEA
44.8
EUR
46801.MD
46801.03
46801.06
Polyethylene glycol diacrylate (PEGDA), M.W. 8,000
Polyethylene glycol diacrylate (PEGDA)
CAS No.: 26570-48-9
Molecular Formula: CH2=COC(OCH2CH2)nOCOC=CH2
Storage Temperature: 2°C to 8°C
Physical Form: Granular solid
Appearance: White to off-white
MDL No.: MFCD00081876
Formel:
CH₂=COC(OCH₂CH₂)nOCOC=CH₂ |
MDL:
MFCD00081876 CAS-Nummer: 26570-48-9 EINECS: 607-960-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...