Chloro(1,5-cyclooctadiene)(pentamethylcyclopentadienyl)ruthenium(II)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
46803.03
46803.03EA
391
EUR
46803.03
Chloro(1,5-cyclooctadiene)(pentamethylcyclopentadienyl)ruthenium(II)
Chloro(1,5-cyclooctadiene)(pentamethylcyclopentadienyl)ruthenium(II)
CAS No.: 92390-26-6
Molecular Formula: C18H27ClRu
Formula Weight: 379.94
Storage Temperature: -30°C to -10°C
Physical Form: Crystalline solid
Appearance: Yellow-brown
Melting Point: 143-147°
UN No.: UN3395
MDL No.: MFCD07369035
Formel:
C₁₈H₂₇ClRu Schmelzpunkt: 143…147 °C |
MDL:
MFCD07369035 CAS-Nummer: 92390-26-6 ADR: 4.3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...