Natriumcarbonat concentrate for dilution to 0.05 mol/l (0.1 N) aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Lieferant: Avantor
Synonyme:
wasserfreie Soda
4683.EA
0
EUR
4683.
Natriumcarbonat concentrate for dilution to 0.05 mol/l (0.1 N) aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Natriumcarbonat
Formel:
Na₂CO₃ Molecular Weight: 105.99 g/mol Siedepunkt: 1600 Schmelzpunkt: 854 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003494 CAS-Nummer: 497-19-8 EINECS: 207-838-8 Merck Index: 14,08596 |
|
Spezifikation Testergebnisse
Titer (N) | 0.099-0.101 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...