Platin ≥99,98% (Metall-Basis), Pulver -200/+400 mesh
Lieferant: Thermo Fisher Scientific
Synonyme:
Platinium
46830.03
46830.06
46830.03EA
309
EUR
46830.03
46830.06
Platin ≥99,98% (Metall-Basis), Pulver -200/+400 mesh
Platinum
Formel:
Pt Molecular Weight: 195.08 g/mol Siedepunkt: 3827 Schmelzpunkt: 1769 Dichte: 21.4 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011179 CAS-Nummer: 7440-06-4 EINECS: 231-116-1 UN: 3089 ADR: 4.1 II Merck Index: 12,07689 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...