Sepabeads® SP207 synthetisch adsorbierendes Kunstharz brominated PS-DVB P.R. 110 angstroms, hoch poröser Typ

Lieferant: Thermo Scientific

46848.A1
46848.A1EA 202 EUR
46848.A1
Sepabeads® SP207 synthetisch adsorbierendes Kunstharz brominated PS-DVB P.R. 110 angstroms, hoch poröser Typ
Sepabeads® SP207

Applications: Bromination of the aromatic ring provides increased hydrophobicity, and consequently, increased selectivity for hydrophobic molecules versus conventional styrene/DVB synthetic adsorbents like Diaion® HP20 resin.

Lagertemperatur: Raumtemperatur
CAS-Nummer: 87924-01-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR