Cefodizime sodium

Lieferant: Thermo Scientific
Gefahr

Synonyme: Sodium (6R,7R)-7-((Z)-2-(2-aminothiazol-4-yl)-2-(methoxyimino)acetamido)-3-(((5-(carboxylatomethyl)-4-methylthiazol-2-yl)thio)methyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate

468680010
468680010.EA 298 EUR
468680010.
Cefodizime sodium
Cefodizime sodium
Formel: C₂₀H₁₈N₆Na₂O₇S₄
Molecular Weight: 628,63 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00897059
CAS-Nummer: 86329-79-5
Merck Index: 13,01941

Bestellen

Spezifikation Testergebnisse

Appearance white to light yellow powder
Infrared spectrum Conforms
HPLC ≥96.0 %
Solubility (1mg/ml in DMSO) Clear colorless solution
Water ≤4 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR