Aminopterin 97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 4-Aminofolic acid, 4-Aminopteroyl-L-glutamic acid, 2-(4-((2,4-Diaminopteridin-6-yl)methylamino)-benzamido)pentanedioic acid, (S)-2-(4-(((2,4-Diaminopteridin-6-yl)methyl)amino)benzamido)pentanedioic acid, 4-Amino-PGA

468751000
468751000.EA 215 EUR
468751000.
Aminopterin 97%
Aminopterin
Formel: C₁₉H₂₀N₈O₅
Molecular Weight: 440,42 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00036692
CAS-Nummer: 54-62-6
EINECS: 200-209-9
UN: 2811
ADR: 6.1,I
Merck Index: 13,00471

Bestellen

Spezifikation Testergebnisse

Appearance light yellow to yellow-brown powder
Infrared spectrum Conforms
HPLC ≥96.0 %
Water ≤10 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR