Meisoindigo

Lieferant: Thermo Scientific

Synonyme: N-Methylisoindigotin, 3-(1,2-Dihydro-2-oxo-3H-indol-3-ylidene)-1,3-dihydro-1-methyl-2H-indol-2-one, 1-Methyl-[3,3'-biindolinylidene]-2,2'-dione

468801000
468801000.EA 413 EUR
468801000.
Meisoindigo
Meisoindigo
Formel: C₁₇H₁₂N₂O₂
Molecular Weight: 276,29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05988503
CAS-Nummer: 97207-47-1

Bestellen

Spezifikation Testergebnisse

Appearance orange to dark red to brown crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR