Brimonidine tartrate 99%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 5-Bromo-N-(4,5-dihydro-1H-imidazol-2-yl)quinoxalin-6-amine (2R,3R)-2,3-dihydroxysuccinate

468842500
468842500.EA 278 EUR
468842500.
Brimonidine tartrate 99%
Brimonidine tartrate
Formel: C₁₅H₁₆BrN₅O₆
Molecular Weight: 442,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07773072
CAS-Nummer: 70359-46-5
UN: 2811
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance white to light yellow to light brown powder
Infrared spectrum Conforms
HPLC ≥98.5 %
Titration with HClO4 99 to 101 %
Loss on drying ≤0.5 % (1 g, 105°C, 3 h)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR