Diiodo(p-cymene)ruthenium(II) dimer, gelb, kristallin

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Di-µ-iodobis[(p-cymene)iodoruthenium(II)]

46886.03 46886.06
46886.03EA 110 EUR
46886.03 46886.06
Diiodo(p-cymene)ruthenium(II) dimer, gelb, kristallin
Diiodo(p-cymene)ruthenium(II) dimer

  • Sensitivity: Moisture and light sensitive
  • Storage: Store under nitrogen


Diiodo(p-cymene)ruthenium(II) dimer can be applied as a catalyst for many reactions such as ruthenium-catalysed hydrogenation, preparation of Walphos-type ferrocenyl-aryl diphosphine ligands, ruthenium-catalysed synthesis reactions, and enantioselective hydrogenation of ketones and olefins.


Insoluble in water.

Formel: C₂₀H₂₈I₄Ru₂
Molecular Weight: 978.20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08064027
CAS-Nummer: 90614-07-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR